Human Recombinant Galectin-3 (from E. coli)
Supplier: Peprotech
450-38-10UG 450-38-50UG 450-38-100UG
10781-362EA 209.84 CAD
10781-362 10781-364 10781-366
Human Recombinant Galectin-3 (from E. coli)
Proteins and Peptides
Lectins, of either plant or animal origin, are carbohydrate-binding proteins that interact with glycoproteins and glycolipids on the surface of animal cells. The Galectins are lectins that recognize and interact with β-galactoside moieties. Galectin-3 regulates a number of biological processes, including embryogenesis, inflammatory responses, cell progression and metastasis. Galectin-3 is normally expressed in epithelia of a variety of tissues, including colon and endometrium, and in various inflammatory cells, including macrophages. Galectin-3 can function intracellularly, controlling the cell cycle and preventing T-cell apoptosis, and also extracellularly, by activating various cells, including monocytes/macrophages, mast cells, neutrophils, and lymphocytes. Expression of Galectin-3 is affected by neoplastic transformation, being up-regulated in certain types of lymphomas, and in thyroid and hepatic carcinomas. Conversely, it is down-regulated in other cancers such as colon, breast, ovarian, and uterine. Recombinant Human Galectin-3 is a globular 26.0 kDa protein containing 250 amino acid residues, but no disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.