Human Recombinant GMF-beta (from E. coli)
Supplier: Peprotech
450-37-10UG 450-37-100UG 450-37-250UG 450-37-500UG 450-37-1MG
10781-352EA 554.79 CAD
10781-352 10781-354 10781-356 10781-358 10781-360
Human Recombinant GMF-beta (from E. coli)
Proteins and Peptides
GMF-β is a brain-specific protein that belongs to the actin-binding proteins ADF structural family. GMF-β appears to play a role in the differentiation, maintenance, and regeneration of the nervous system. It also supports the progression of certain auto-immune diseases, possibly through its ability to induce the production and secretion of various pro-inflammatory cytokines. Recombinant Human GMF-β is a 16.5 kDa globular protein containing 141 amino acid residues, including one intramolecular disulfide bond.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.