Human Recombinant NT-4 (from E. coli)
Supplier: Peprotech
450-04-2UG 450-04-10UG 450-04-50UG 450-04-100UG 450-04-250UG 450-04-500UG 450-04-1MG
10781-180EA 221.22 CAD
10781-180 10781-182 10781-184 10781-186 10781-188 10781-190 10781-192
Human Recombinant NT-4 (from E. coli)
Proteins and Peptides
NT-4 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-3. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-4 is expressed in the prostate, thymus, placenta, and skeletal muscle. NT-4 can signal through the LNGFR and trkB receptors, and promotes the survival of peripheral sensory sympathetic neurons. Recombinant Human NT-4 is a noncovalently linked homodimer of two 14.0 kDa polypeptide monomers (260 total amino acid residues).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.