Human Recombinant TIMP-1 (from E. coli)
Supplier: Peprotech
410-01-2UG 410-01-10UG 410-01-50UG 410-01-100UG 410-01-250UG 410-01-500UG 410-01-1MG
10781-078EA 209.84 CAD
10781-078 10781-080 10781-082 10781-084 10781-086 10781-088 10781-090
Human Recombinant TIMP-1 (from E. coli)
Proteins and Peptides
TIMP-1 is an extracellular inhibitor of MMPs, including MMP-1, -2, -3, -7, -8, -9, -10, -11, -12, -13, and -16. It belongs to the I35 (TIMP) family of irreversible protease inhibitors that function as key modulators of extracellular matrix degradation during tissue development and remodeling. TIMP-1 can also act through an MMP-independent mechanism to promote erythropoiesis by stimulating proliferation and differentiation of erythroid progenitors. Recombinant Human TIMP-1 is a 20.6 kDa protein containing 184 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.