Rat Recombinant G-CSF (from E. coli)
Supplier: Peprotech
400-37-2UG 400-37-10UG 400-37-50UG 400-37-100UG 400-37-250UG 400-37-500UG 400-37-1MG
10781-052EA 227.33 CAD
10781-052 10781-054 10781-056 10781-058 10781-060 10781-062 10781-064
Rat Recombinant G-CSF (from E. coli)
Proteins and Peptides
G-CSF is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It is produced in response to specific stimulation by a variety of cells, including macrophages, fibroblasts, endothelial cells and bone marrow stroma. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Recombinant Rat G-CSF is a 21.6 kDa protein consisting of 196 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.