Rat Recombinant Resistin (from E. coli)
Supplier: Peprotech
400-35-5UG 400-35-25UG 400-35-100UG 400-35-250UG 400-35-500UG 400-35-1MG
10781-028EA 209.84 CAD
10781-028 10781-030 10781-032 10781-034 10781-036 10781-038
Rat Recombinant Resistin (from E. coli)
Proteins and Peptides
Resistin belongs to a family of tissue-specific cytokines termed FIZZ (found in inflammatory zones) and RELM. The four known members of this family, resistin, RELMα, RELMβ, and RELMγ, share a highly conserved C-terminal domain, characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. Resistin is an adipose-derived cytokine (adipokine) whose physiological function and molecular targets are largely unknown. Studies have shown that resistin suppresses insulin's ability to stimulate glucose uptake, and postulated that resistin might be an important link between obesity and Type 2 diabetes. Other studies have indicated that resistin expression is severely suppressed in obesity, and that it may act as a feedback regulator of Adipogenesis. Recombinant Rat Resistin is a 20.0 kDa, disulfide-linked, homodimeric protein composed of two 94 identical amino acid chains linked by a single disulfide bond.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.