Rat Recombinant IP-10 (from E. coli)
Supplier: Peprotech
IP-10 is a CXC chemokine that signals through the CXCR3 receptor. IP-10 selectively chemoattracts Th1 lymphocytes and monocytes, and inhibits cytokine-stimulated hematopoietic progenitor cell proliferation. Additionally, it is angiostatic and mitogenic for vascular smooth muscle cells. Recombinant Rat IP-10 (CXCL10) is an 8.6 kDa protein consisting of 77 amino acids, including the four conserved cysteine residues present in CXC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.