Rat Recombinant GRO-beta/MIP-2 (from E. coli)
Supplier: Peprotech
400-11-5UG
400-11-25UG
400-11-100UG
400-11-250UG
400-11-500UG
400-11-1MG
10780-974EA
221.22
CAD
10780-974
10780-976
10780-978
10780-980
10780-982
10780-984
Rat Recombinant GRO-beta/MIP-2 (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. Recombinant Rat GRO-β/MIP-2 (CXCL2) is a 7.9 kDa protein consisting of 73 amino acids, including the ‘ELR’ motif common to the CXC chemokine family that binds to CXCR1 or CXCR2.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.