Rat Recombinant IL-4 (from E. coli)
Supplier: Peprotech
400-04-5UG 400-04-20UG 400-04-50UG 400-04-100UG 400-04-250UG 400-04-500UG 400-04-1MG
10780-896EA 223.76 CAD
10780-896 10780-898 10780-900 10780-902 10780-904 10780-906 10780-908
Rat Recombinant IL-4 (from E. coli)
Proteins and Peptides
IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival and gene expression. Produced by mast cells, T cells and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. Recombinant Rat IL-4 is a 14.0 kDa globular protein containing 126 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.