Mouse Recombinant IFN-gamma (from E. coli)
Supplier: Peprotech
315-05-20UG 315-05-100UG 315-05-250UG 315-05-500UG
10780-720EA 223.89 CAD
10780-720 10780-722 10780-724 10780-726
Mouse Recombinant IFN-gamma (from E. coli)
Proteins and Peptides
IFN-γ is an acid-labile interferon produced by CD4 and CD8 T lymphocytes, as well as activated NK cells. IFN-γ receptors are present in most immune cells, which respond to IFN-γ signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-γ signaling in antigen-presenting, cells and antigen-recognizing B and T lymphocytes, regulates the antigen-specific phases of the immune response. Additionally, IFN-γ stimulates a number of lymphoid cell functions, including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Human IFN-γ is species-specific and is biologically active only in human and primate cells. Recombinant Murine IFN-γ is a 15.6 kDa protein containing 134 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.