Mouse Recombinant M-CSF (from E. coli)
Supplier: Peprotech
315-02-2UG 315-02-10UG 315-02-50UG 315-02-100UG 315-02-250UG 315-02-500UG 315-02-1MG
10780-692EA 241.19 CAD
10780-692 10780-694 10780-696 10780-698 10780-700 10780-702 10780-704
Mouse Recombinant M-CSF (from E. coli)
Proteins and Peptides
M-CSF is a potent hematopoietic factor produced by a variety of cells, including lymphocytes, monocytes, fibroblasts, endothelial cells, myoblasts and osteoblasts. It is a key regulator of cellular proliferation, differentiation, and survival for blood monocytes, tissue macrophages, and their respective progenitor cells. M-CSF has been shown to play important roles in modulating dermal thickness and fertility. M-CSF is clinically used in the treatment of infection, malignancies and atherosclerosis. It facilitates hematopoietic recovery after bone marrow transplantation. Human M-CSF is reactive in murine systems, but the murine molecule exhibits no activity on human cells. Recombinant Murine M-CSF is a 36.4 kDa homodimeric protein consisting of two 156 amino acid polypeptide subunits.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.