Human Recombinant MEC (from E. coli)
Supplier: Peprotech
300-57-5UG 300-57-20UG
10780-620EA 209.84 CAD
10780-620 10780-622
Human Recombinant MEC (from E. coli)
Proteins and Peptides
MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor, and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines. Recombinant Human MEC (CCL28) is a 12.3 kDa protein containing 108 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.