Human Recombinant LD78beta (from E. coli)
Supplier: Peprotech
300-56-5UG 300-56-20UG 300-56-100UG 300-56-250UG 300-56-500UG 300-56-1MG
10780-608EA 209.84 CAD
10780-608 10780-610 10780-612 10780-614 10780-616 10780-618
Human Recombinant LD78beta (from E. coli)
Proteins and Peptides
LD78β is a CC chemokine that is closely related to MIP-1α. It signals through the CCR5 receptor and the beta-chemokine receptor, D6. LD78β has been shown to exhibit potent activity in HIV suppression assays. Recombinant Human LD78β (CCL3L1) is a 7.7 kDa protein containing 70 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.