Human Recombinant CTACK (from E. coli)
Supplier: Peprotech
300-54-5UG 300-54-20UG 300-54-100UG 300-54-250UG 300-54-500UG 300-54-1MG
10780-584EA 209.84 CAD
10780-584 10780-586 10780-588 10780-590 10780-592 10780-594
Human Recombinant CTACK (from E. coli)
Proteins and Peptides
CTACK is a keratinocyte-derived CC chemokine which signals through the CCR10 receptor. Both CTACK and CCR10 are expressed in normal and irritated epithelial cells. CTACK selectively attracts CLA+ T-cells and directs them into the skin. CTACK contains the four highly conserved cysteine residues present in most CC chemokines. The mature protein contains 88 amino acid residues. Recombinant Human CTACK (CCL27) is a 10.2 kDa protein containing 88 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.