Human Recombinant Apo-SAA (from E. coli)

Supplier: Peprotech

300-13-100UG 300-13-250UG 300-13-500UG 300-13-1MG
10780-480EA 561.58 CAD
10780-480 10780-482 10780-484 10780-486
Human Recombinant Apo-SAA (from E. coli)
Proteins and Peptides

Human Apo-SAA is a 104 amino acid polypeptide that circulates primarily in association with high-density lipoproteins (HDL). The level of Apo-SAA, normally 1-5 μg/ml in plasma, increases 500-1000 fold within 24 hours of an inflammatory stimulus and, under these conditions, is the most abundant HDL apolipoprotein. The human SAA gene codes for a 122 amino acid polypeptide, which contains an 18 amino acid N-terminal signal sequence. Recombinant Apo-SAA is a consensus SAA molecule corresponding to human Apo-SAA1α, except for the presence of an N-terminal methionine, the substitution of asparagine for aspartic acid at position 60, and arginine for histidine at position 71 (the latter two substituted residues are present in Apo-SAA2β). The calculated molecular weight of Recombinant Human Apo-SAA is 11.7 kDa.


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name Apo-SAA
Environmentally Preferable
Protein/peptide type Recombinant
Protein Function Cytokines and Growth Factors
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID 6288 P0DJ18
Sequence MRSFFSFLGE AFDGARDMWR AYSDMREANY IGSDKYFHAR GNYDAAKRGP GGVWAAEAIS NARENIQRFF GRGAEDSLAD QAANEWGRSG KDPNHFRPAG LPEKY
Protease-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.98
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 11.7 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR