Human Recombinant MIP-1alpha (from E. coli)
Supplier: Peprotech
300-08-5UG 300-08-20UG 300-08-50UG 300-08-100UG 300-08-250UG 300-08-500UG 300-08-1MG
10780-462EA 221.22 CAD
10780-462 10780-464 10780-466 10780-468 10780-470 10780-472 10780-474
Human Recombinant MIP-1alpha (from E. coli)
Proteins and Peptides
Both MIP-1α and MIP-1β are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens, by regulating the trafficking, and activation state, of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1α and MIP-1β exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1α selectively attracting CD8+ lymphocytes, and MIP-1β selectively attracting CD4+ lymphocytes. Additionally, MIP-1α and MIP-1β have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1α and MIP-1β are active on human and murine hematopoietic cells. Recombinant Human MIP-1α (CCL3) is a 7.8 kDa protein containing 70 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.