Human Recombinant LIF (from E. coli)

Supplier: Peprotech

300-05-5UG 300-05-25UG 300-05-100UG 300-05-250UG 300-05-500UG 300-05-1MG
10780-422EA 223.16 CAD
10780-422 10780-424 10780-426 10780-428 10780-430 10780-432
Human Recombinant LIF (from E. coli)
Proteins and Peptides

LIF is a pleiotrophic factor produced by multiple cell types, including T cells, myelomonocytic lineages, fibroblasts, liver, heart and melanoma. LIF promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. Other activities include the stimulation of acute phase protein synthesis by hepatocytes, stimulation of differentiation of cholinergic nerves, and suppression of adipogenesis by inhibiting the lipoprotein lipase in adipocytes. While human LIF is active on mouse cells and is widely used in the maintenance of murine ESC to prevent spontaneous differentiation, mouse LIF is not active on human cells due to its inability to bind to the human LIF receptor. Recombinant Human LIF is a 19.6 kDa protein containing 180 amino acids residues, including three disulfide bonds.


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name LIF
Protein synonyms Leukemia Inhibitory Factor|Differentiation-stimulating factor|D factor|Melanoma-derived LPL inhibitor (MLPLI)
Environmentally Preferable
Protein/peptide type Recombinant
Protein Function Cytokines and Growth Factors
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID 3976 P15018
Sequence SPLPITPVNA TCAIRHPCHN NLMNQIRSQL AQLNGSANAL FILYYTAQGE PFPNNLDKLC GPNVTDFPPF HANGTEKAKL VELYRIVVYL GTSLGNITRD QKILNPSALS LHSKLNATAD ILRGLLSNVL CRLCSKYHVG HVDVTYGPDT SGKDVFQKKK LGCQLLGKYK QIIAVLAQAF
Protease-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.98
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 19.6 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR