Human Recombinant GM-CSF (from E. coli)
Supplier: Peprotech
300-03-5UG 300-03-20UG 300-03-50UG 300-03-100UG 300-03-250UG 300-03-500UG 300-03-1MG
10780-396EA 234.54 CAD
10780-396 10780-398 10780-400 10780-402 10780-404 10780-406 10780-408
Human Recombinant GM-CSF (from E. coli)
Proteins and Peptides
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages, and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. It is produced in endothelial cells, monocytes, fibroblasts and T lymphocytes. GM-CSF inhibits neutrophil migration and enhances the functional activity of the mature end-cells. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant Human GM-CSF is a 14.6 kDa globular protein consisting of 128 amino acids, containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.