Human Recombinant IFN-?1 (from E. coli)
Supplier: Peprotech
300-02L-5UG 300-02L-20UG 300-02L-100UG 300-02L-250UG 300-02L-500UG 300-02L-1MG
10780-384EA 220.96 CAD
10780-384 10780-386 10780-388 10780-390 10780-392 10780-394
Human Recombinant IFN-?1 (from E. coli)
Proteins and Peptides
IFN λ1, 2, and 3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family and the interferons. All three IFN-lambdas use a distinct receptor system composed of an IFN-λR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defenses similar to, but distinct from, that of type I interferons. Recombinant Human IFN-λ1 is a 19.8 kDa protein containing 178 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.