Mouse Recombinant SDF-1alpha (from E. coli)
Supplier: PeproTech
250-20A-100UG 250-20A-10UG 250-20A-2UG 250-20A-50UG
10780-296EA 213.44 CAD
10780-296 10780-298 10780-300 10780-302
Mouse Recombinant SDF-1alpha (from E. coli)
Proteins and Peptides
SDF-1α and β are stromal-derived, CXC chemokines that signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV-suppressive activity in cells expressing the CXCR4 receptor. Human and murine SDF-1 proteins act across species. SDF-1α and β contain the four highly conserved cysteine residues present in CXC chemokines. The mature SDF-1α protein is the result of alternative splicing of the SDF-1 gene and contains 68 amino acid residues. Recombinant Murine SDF-1α (CXCL12) is a 7.9 kDa protein containing 68 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.