Mouse Recombinant IL-5 (from E. coli)
Supplier: Peprotech
215-15-5UG 215-15-25UG 215-15-50UG 215-15-100UG 215-15-250UG 215-15-500UG 215-15-1MG
10780-154EA 229.35 CAD
10780-154 10780-156 10780-158 10780-160 10780-162 10780-164 10780-166
Mouse Recombinant IL-5 (from E. coli)
Proteins and Peptides
IL-5 is a hematopoietic growth factor that stimulates the proliferation and activation of eosinophils. Produced by mast cells, T cells, and eosinophils, IL-5 plays an important role in inducing cell-mediated immunity against parasitic infections and certain tumors. Elevated levels of IL-5 lead to Eosinophilia, which may result in the induction of asthma and other allergic diseases. Human and murine IL-5 is cross-species reactive. Recombinant Murine IL-5 is a 26.2 kDa disulfide-linked homodimeric protein containing two 113 amino acid chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.