Mouse Recombinant IL-3 (from E. coli)
Supplier: Peprotech
213-13-2UG AF-213-13-10UG 213-13-50UG 213-13-100UG 213-13-250UG 213-13-500UG 213-13-1MG
10780-126EA 247.25 CAD
10780-126 10780-128 10780-130 10780-132 10780-134 10780-136 10780-138
Mouse Recombinant IL-3 (from E. coli)
Proteins and Peptides
IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoiesis, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is a species-specific, variably glycosylated cytokine. Recombinant Murine IL-3 is a 15.1 kDa globular protein containing 135 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.