Mouse Recombinant IL-1alpha (from E. coli)
Supplier: Peprotech
211-11A-2UG 211-11A-10UG 211-11A-50UG 211-11A-100UG 211-11A-250UG 211-11A-500UG 211-11A-1MG
10780-084EA 226.59 CAD
10780-084 10780-086 10780-088 10780-090 10780-092 10780-094 10780-096
Mouse Recombinant IL-1alpha (from E. coli)
Proteins and Peptides
IL-1α is a non-secreted, proinflammatory cytokine produced in a variety of cells, including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1α and IL-1β bind to the same receptor and have similar, if not identical, biological properties. These cytokines have a broad range of activities including the stimulation of thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity, and the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. Recombinant Murine IL-1α is a 17.9 kDa protein containing 156 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.