Mouse Recombinant IL-33 (from E. coli)
Supplier: Peprotech
210-33-2UG 210-33-10UG 210-33-50UG 210-33-100UG 210-33-250UG 210-33-500UG 210-33-1MG
10780-070EA 220.96 CAD
10780-070 10780-072 10780-074 10780-076 10780-078 10780-080 10780-082
Mouse Recombinant IL-33 (from E. coli)
Proteins and Peptides
IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It binds and signals through the IL-1RL1/ST2 receptor, to activate NF-κB and MAP kinases. IL-33 induces production of TH2 cell related cytokines, including IL-4, IL-5 and IL-13, and exerts multiple inflammation related bioactivities. Recombinant Murine IL-33 is a 17.5 kDa protein containing 158 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.