Mouse Recombinant SF-20 (from E. coli)
Supplier: Peprotech
210-25-2UG 210-25-10UG 210-25-100UG 210-25-250UG 210-25-500UG 210-25-1MG
10780-046EA 221.22 CAD
10780-046 10780-048 10780-050 10780-052 10780-054 10780-056
Mouse Recombinant SF-20 (from E. coli)
Proteins and Peptides
Murine SF-20 is a bone marrow stroma-derived growth factor. SF20 is expressed in bone marrow, spleen stroma cells, resting mononuclear cells, resting CD8+ and CD19+ cells, and activated CD8+ T cells, and has been shown to bind to the surface of cells expressing the receptor TSA-1 (Thymic shared Ag-1). Among its biological activities, SF20 stimulates the proliferation of FDCP2 cells (a mouse factor-dependent hemopoietic cell line) and mouse lymphoid cells. Recombinant Murine SF-20 is a 15.8 kDa protein containing 143 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.