Mouse Recombinant IL-17F (from E. coli)
Supplier: Peprotech
210-17F-5UG
210-17F-25UG
210-17F-50UG
210-17F-100UG
210-17F-250UG
210-17F-500UG
210-17F-1MG
10780-004EA
209.84
CAD
10780-004
10780-006
10780-008
10780-010
10780-012
10780-014
10780-016
Mouse Recombinant IL-17F (from E. coli)
Proteins and Peptides
IL-17F, a member of the IL-17 family of structurally related cytokines, has been shown to stimulate proliferation and activation of T-cells and PBMCs. IL-17F also regulates cartilage matrix turnover and inhibits angiogenesis. Recombinant Murine IL-17F is a 30.0 kDa, disulfide-linked homodimer consisting of two 134 amino acid residue chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.