Human Recombinant IL-17B (from E. coli)

Supplier: Peprotech

200-28-5UG 200-28-25UG 200-28-100UG 200-28-250UG 200-28-500UG 200-28-1MG
10779-816EA 209.84 CAD
10779-816 10779-818 10779-820 10779-822 10779-824 10779-826
Human Recombinant IL-17B (from E. coli)
Proteins and Peptides

IL-17B is a disulfide-linked homodimer of two 161 amino acid polypeptide chains. It belongs to the IL-17 family of structurally-related cytokines that share a highly conserved C-terminal region, but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17B is expressed by T-cells, and has been shown to stimulate release of TNF-α and IL-1β from cells of the monocyte lineage. Recombinant Human IL-17B is a 36.6 kDa non-disulfide-linked homodimer of two 161 amino acid polypeptide chains.


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name IL-17B
Protein synonyms Cytokine ZCYTO7|NIRF|IL-20|Interleukin-17B
Environmentally Preferable
Protein/peptide type Recombinant
Protein Function Cytokines and Growth Factors
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID 27190 Q9HF5
Sequence MQPRSPKSKR KGQGRPGPLA PGPHQVPLDL VSRMKPYARM EEYERNIEEM VAQLRNSSEL AQRKCEVNLQ LWMSNKRSLS PWGYSINHDP SRIPVDLPEA RCLCLGCVNP FTMQEDRSMV SVPVFSQVPV RRRLCPPPPR TGPCRQRAVM ETIAVGCTCI F
Protease-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.98
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 36.6 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR