Human Recombinant IL-17D (from E. coli)
Supplier: Peprotech
200-27-5UG 200-27-25UG 200-27-100UG 200-27-250UG 200-27-500UG 200-27-1MG
10779-804EA 209.84 CAD
10779-804 10779-806 10779-808 10779-810 10779-812 10779-814
Human Recombinant IL-17D (from E. coli)
Proteins and Peptides
IL-17D is a disulfide-linked homodimer of two 185 amino acid polypeptide chains. It belongs to the IL-17 family of structurally-related cytokines that share a highly conserved C-terminal region, but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17D has the ability to stimulate the production of IL-6, IL-8 and GM-CSF, and inhibits hemopoiesis of myeloid progenitor cells in colony-forming assays. Recombinant Human IL-17D is a 40.5 kDa disulfide-linked homodimer of two 185 amino acid polypeptide chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.