Human Recombinant IL-23 (from (BTI-Tn-5B1-4) Hi-5 Insect Cells)
Supplier: Peprotech
200-23-2UG 200-23-10UG 200-23-50UG 200-23-100UG 200-23-250UG 200-23-500UG 200-23-1MG
10779-764EA 226.59 CAD
10779-764 10779-766 10779-768 10779-770 10779-772 10779-774 10779-776
Human Recombinant IL-23 (from (BTI-Tn-5B1-4) Hi-5 Insect Cells)
Proteins and Peptides
IL-23 is a proinflammatory, heterodimeric protein composed of two subunits: a unique p19 subunit, and a p40 subunit that is shared with IL-12. IL-23 is secreted by activated dendritic cells and macrophages, and signals though a receptor comprised of IL-23R complexed with IL-12Rβ2. IL-23 has been shown to enhance proliferation of memory T cells. It also stimulates the production of IFN-γ in NK cells, induces IL-17 production, and drives Th17-mediated responses. Recombinant Human IL-23 is a 53.5 kDa, heterodimeric protein consisting of the two subunits, p19 (170 amino acids) and p40 (306 amino acids).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.