Human Recombinant IL-19 (from E. coli)
Supplier: Peprotech
200-19-2UG 200-19-10UG 200-19-100UG 200-19-250UG 200-19-500UG 200-19-1MG
10779-712EA 209.84 CAD
10779-712 10779-714 10779-716 10779-718 10779-720 10779-722
Human Recombinant IL-19 (from E. coli)
Proteins and Peptides
IL-19 belongs to the IL-10 family of regulatory cytokines, which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences, but they are dissimilar in their biological functions. Preliminary data suggests that IL-19 is a proinflammatory cytokine, because it up-regulates IL-6 and TNF-α, and induces apoptosis through TNF-α. IL-19 signals through the type I IL-20R. Human and murine IL-19 share 71% amino acid sequence identity. Recombinant Human IL-19 is a 35.8 kDa homodimer of two 154 amino acid chains. In solution, IL-19 exists predominantly as a non-disulfide-linked dimer.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.