Human Recombinant IL-17A (from E. coli)
Supplier: Peprotech
200-17-5UG 200-17-25UG 200-17-50UG 200-17-100UG 200-17-250UG 200-17-500UG 200-17-1MG
10779-698EA 223.76 CAD
10779-698 10779-700 10779-702 10779-704 10779-706 10779-708 10779-710
Human Recombinant IL-17A (from E. coli)
Proteins and Peptides
The originally described IL-17 protein, now known as IL-17A, is a homodimer of two 136 amino acid chains that are secreted by activated T-cells, which act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region, but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells. Recombinant Human IL-17A is a 31.3 kDa, disulfide-linked homodimer of two 137 amino acid, polypeptide chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.