Human Recombinant IL-8 (from E. coli)
Supplier: Peprotech
200-08-5UG 200-08-25UG 200-08-100UG 200-08-250UG 200-08-500UG
10779-686EA 221.03 CAD
10779-686 10779-688 10779-690 10779-692 10779-694
Human Recombinant IL-8 (from E. coli)
Proteins and Peptides
IL-8 is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils. Recombinant Human IL-8 (CXCL9) (endothelial-derived) is an 8.9 kDa protein containing 77 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.