Human Recombinant IL-6 (from E. coli)
Supplier: Peprotech
200-06-5UG 200-06-20UG 200-06-50UG 200-06-100UG 200-06-250UG 200-06-500UG 200-06-1MG
10779-644EA 238.19 CAD
10779-644 10779-646 10779-648 10779-650 10779-652 10779-654 10779-656
Human Recombinant IL-6 (from E. coli)
Proteins and Peptides
IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses.
  • ecombinant Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues
Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6Rα and gp130. Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells.
Caution: For research use only. Not for use in diagnostic or therapeutic procedures.