Mycoplasma Recombinant Mycoplasma Arginine Deiminase (from E. coli)
Supplier: Peprotech
150-12-5UG 150-12-20UG 150-12-100UG 150-12-250UG 150-12-500UG 150-12-1MG
10779-524EA 221.22 CAD
10779-524 10779-526 10779-528 10779-530 10779-532 10779-534
Mycoplasma Recombinant Mycoplasma Arginine Deiminase (from E. coli)
Enzymes
Arginine Deiminase (ADI) is a microbial enzyme from Mycoplasma produced in E.coli. It has high affinity to L-arginine and hydrolyzes L-arginine to citrulline and ammonia. Low concentrations of ADI have been shown to inhibit proliferation in certain cultured cells by arresting the cell cycle in G Recombinant Mycoplasma Arginine Deiminase is a 46.3 kDa protein consisting of 409 amino acids.
  • Origin: USA
  • Accession number: P23793
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.