Human Recombinant Endostatin (from E. coli)
Supplier: Peprotech
150-01-20UG 150-01-100UG 150-01-250UG 150-01-500UG 150-01-1MG
10779-466EA 209.84 CAD
10779-466 10779-468 10779-470 10779-472 10779-474
Human Recombinant Endostatin (from E. coli)
Proteins and Peptides
Endostatin is a naturally occurring 20 kDa polypeptide derived from the C-terminal portion of type XVIII collagen. It functions as an anti-angiogenic cytokine that is expressed in various organs with the highest levels in liver, lung and kidney. Endostatin inhibits angiogenesis by blocking the pro-angiogenic activities of VEGF and FGF-basic. Recombinant Human Endostatin is a 20.2 kDa protein consisting of 184 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.