Human Recombinant Vitronectin (from HEK293 Cells)
Supplier: Peprotech
AF-140-09-100UG AF-140-09-500UG
10779-426EA 279.99 CAD
10779-426 10779-428 10779-430
Human Recombinant Vitronectin (from HEK293 Cells)
Proteins and Peptides
Vitronectin is a secreted glycoprotein that is synthesized in the liver. It circulates primarily in monomeric form, but can undergo conformational change to a structure that forms disulfide-linked multimers. The multimeric vitronectin can efficiently bind to, and incorporate into, the extracellular matrix. Within the matrix, vitronectin can support cell adhesion through binding to various integrins and other proteoglycans. Additionally, recombinant vitronectin can function as a chemically-defined matrix component in human embryonic stem cell renewal media. Recombinant Human Vitronectin is a 459 amino acid, single-chain, monomeric protein, which migrates at an apparent molecular weight of 75 kDa by SDS-PAGE under reducing conditions. The calculated molecular weight of Recombinant Human Vitronectin is 52.2 kDa.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.