Human Recombinant INSL5/INSL7 (from E. coli)
Supplier: Peprotech
130-05-5UG 130-05-25UG 130-05-100UG 130-05-250UG 130-05-500UG 130-05-1MG
10779-348EA 210.25 CAD
10779-348 10779-350 10779-352 10779-354 10779-356 10779-358
Human Recombinant INSL5/INSL7 (from E. coli)
Proteins and Peptides
The Insulin/Relaxin structural family of proteins consists of at least seven members, each containing an insulin-like heterodimeric structure of two polypeptide chains held together by two inter-chain disulfide bonds. The INSL5/INSL7 hybrid contains the β-chain of INSL7 (Relaxin-3), linked to the α-chain of INSL5. The INSL5/INSL7 hybrid protein can specifically activate the G protein-coupled receptors, GPCR135 and GPCR142, but unlike Relaxin-3, does not induce significant activation of the LGR7 receptor. Recombinant Human INSL5/INSL7 Hybrid is a heterodimeric, 5.2 kDa protein consisting of a 27 amino acid β-chain linked to a 21 amino acid α-chain by two disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.