Human Recombinant EPO (from CHO cells)
Supplier: Peprotech
100-64-10UG 100-64-50UG 100-64-100UG 100-64-250UG 100-64-500UG 100-64-1MG
10779-050EA 225.99 CAD
10779-050 10779-052 10779-054 10779-056 10779-058 10779-060
Human Recombinant EPO (from CHO cells)
Proteins and Peptides
Erythropoietin (EPO) is a glycoprotein hormone that is principally known for its role in erythropoiesis, where it is responsible for stimulating proliferation and differentiation of erythroid progenitor cells. The differentiation of CFU-E (Colony Forming Unit-Erythroid) cells into erythrocytes can only be accomplished in the presence of EPO. Physiological levels of EPO in adult mammals are maintained primarily by the kidneys, whereas levels in fetal or neonatal mammals are maintained by the liver. EPO also can exert various non-hematopoietic activities, including vascularization and proliferation of smooth muscle, neural protection during hypoxia, and stimulation of certain B cells. PeproTech’s Human EPO contains 166 amino acid residues and has a calculated molecular weight of approximately 18.4 kDa. As a result of glycosylation, Recombinant Human EPO migrates with an apparent molecular mass of 37.0 kDa by SDS-PAGE gel, under reducing and non-reducing conditions.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.