Human Recombinant Thrombomodulin (from HEK293 Cells)
Supplier: Peprotech
100-58-500UG 100-58-1MG
10779-032EA 5687.97 CAD
10779-032 10779-034
Human Recombinant Thrombomodulin (from HEK293 Cells)
Proteins and Peptides
Thrombomodulin (TM, CD141, THBD) is an endothelial cell-expressed, transmembrane glycoprotein that can form a complex with the coagulation factor, thrombin. The thrombomodulin/thrombin complex converts protein C to its activated form, protein Ca, which in turn proteolytically cleaves and deactivates factor Va and factor VIIIa, two essential components of the coagulation mechanism. This inactivation reduces the generation of additional thrombin, and thereby effectively prevents continued coagulation. Reduced levels of thrombomodulin can correlate with the pathogenesis of certain cardiovascular diseases, such as atherosclerosis and thrombosis. However, the serum levels of the truncated circulating form of thrombomodulin are typically elevated during inflammation and in the presence of various inflammatory-related diseases. The thrombomodulin protein contains 575 amino acids, including an 18 a.a. signal sequence, a 497 a.a. extracellular domain, a 24 a.a. transmembrane sequence, and a 36 a.a. cytoplasmic region. Recombinant Human Thrombomodulin is a 51.4 kDa, 491-amino-acid length glycoprotein containing the extracellular domain of thrombomodulin.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.