Human Recombinant FGF-20 (from E. coli)
Supplier: Peprotech
100-41-3UG 100-41-15UG 100-41-100UG 100-41-250UG 100-41-500UG 100-41-1MG
10778-978EA 209.84 CAD
10778-978 10778-980 10778-982 10778-984 10778-986 10778-988
Human Recombinant FGF-20 (from E. coli)
Proteins and Peptides
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through the FGFR 2c and 3c, and is expressed during limb and brain development. Recombinant Human FGF-20 is a 23.2 kDa protein containing 209 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.