Human Recombinant TGF-alpha (from E. coli)
Supplier: Peprotech
100-16A-20UG 100-16A-100UG 100-16A-250UG 100-16A-500UG 100-16A-1MG
10778-876EA 235.91 CAD
10778-876 10778-878 10778-880 10778-882 10778-884
Human Recombinant TGF-alpha (from E. coli)
Proteins and Peptides
TGF-α is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-α induces anchorage-independent transformation in cultured cells. Human, murine and rat TGF-α are cross-species reactive. Recombinant Human TGF-α is a 50 amino acid polypeptide (5.5 kDa), which shares approximately 40% sequence homology with EGF, including 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.