Mouse Recombinant GMCSF (from E. coli)

Supplier: Peprotech

AF-315-03-5UG AF-315-03-20UG AF-315-03-50UG AF-315-03-100UG AF-315-03-250UG AF-315-03-500UG AF-315-03-1MG
10778-812EA 286.72 CAD
10778-812 10778-814 10778-816 10778-818 10778-820 10778-822 10778-824
Mouse Recombinant GMCSF (from E. coli)
Proteins and Peptides

GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages, and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. It is produced in endothelial cells, monocytes, fibroblasts and T-lymphocytes. GM-CSF inhibits neutrophil migration and enhances the functional activity of the mature end-cells. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant Murine GM-CSF is a 14.2 kDa globular protein consisting of 125 amino acid residues.Manufactured using all Animal-Free reagents.


  • Carrier free
  • Animal-free


MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK


Verified by N-terminal and Mass Spectrometry analyses (when applicable).


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name GMCSF
Environmentally Preferable
Protein/peptide type Recombinant
Species Mouse
Source E. coli
Gene ID 12981
Animal-free Y
Endotoxin content <0.1 ng/μg of protein (<1 EU/μg).
Biological activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine FDC-P1 cells is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 107 units/mg.
Purity Greater than 98% by SDS-PAGE gel and HPLC analyses.
Formulation Lyophilized
Storage conditions −20 °C
Shipping temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR