Human Recombinant G-CSF (from E. coli)
Supplier: Peprotech
AF-300-23-2UG AF-300-23-10UG AF-300-23-100UG AF-300-23-250UG AF-300-23-500UG AF-300-23-1MG
10778-678EA 300.93 CAD
10778-678 10778-680 10778-682 10778-684 10778-686 10778-688
Human Recombinant G-CSF (from E. coli)
Proteins and Peptides
G-CSF is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It is produced in response to specific stimulation by a variety of cells, including macrophages, fibroblasts, endothelial cells and bone marrow stroma. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Human and murine G-CSF are cross-species reactive. Recombinant Human G-CSF is an 18.7 kDa protein consisting of 174 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.