Human Recombinant IFN-beta (from E. coli)
Supplier: Peprotech
AF-300-02B-5UG AF-300-02B-20UG AF-300-02B-100UG AF-300-02B-250UG AF-300-02B-500UG AF-300-02B-1MG
10778-536EA 291.78 CAD
10778-536 10778-538 10778-540 10778-542 10778-544 10778-546
Human Recombinant IFN-beta (from E. coli)
Proteins and Peptides
Proteins of this family play an important role in inducing non-specific resistance against a broad range of viral infections. They also affect cell proliferation and modulate immune responses. Produced by peripheral blood leukocytes and lymphoblastoid cells, IFN-α is an acid-stable molecule that signals through IFN-α/βR, which is also used by IFN-β. Both IFNs have similar anti-viral activity and regulate expression of MHC class I antigens. IFN-α contains four highly conserved cysteine residues that form two disulfide bonds, one of which is necessary for biological activity. PeproTech’s Recombinant Human IFN-β is a 20.0 kDa protein containing 166 amino acid residues. Due to glycosylation, IFN-β has an approximate molecular weight of 22.3 kDa based on SDS-PAGE gel and Mass Spectrometry.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.