Human Recombinant TNF-alpha (from E. coli)
Supplier: Peprotech
AF-300-01A-10UG AF-300-01A-50UG AF-300-01A-100UG AF-300-01A-250UG AF-300-01A-500UG AF-300-01A-1MG
10778-514EA 310.15 CAD
10778-514 10778-516 10778-518 10778-520 10778-522 10778-524
Human Recombinant TNF-alpha (from E. coli)
Proteins and Peptides
TNF-α is a pleiotropic pro-inflammatory cytokine secreted by various cells, including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. It belongs to the TNF family of ligands, and signals through two receptors, TNFR1 and TNFR2. TNF-α is cytotoxic to a wide variety of tumor cells, and is an essential factor in mediating the immune response against bacterial infections. TNF-α also plays a role in the induction of septic shock, autoimmune diseases, rheumatoid arthritis, inflammation, and diabetes. Human and murine TNF-α demonstrate significant cross-species reactivity. TNF-α exists in two forms; a type II transmembrane protein, and a mature soluble protein. The TNF-α transmembrane protein is proteolitically cleaved to yield a soluble, biologically active, 17 kDa TNF-α, which forms a non-covalently linked homotrimer in solution. Recombinant Human TNF-α is a soluble 157 amino acid protein (17.4 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.