Mouse Recombinant IL-6 (from E. coli)
Supplier: Peprotech
AF-216-16-2UG AF-216-16-10UG AF-216-16-50UG AF-216-16-100UG AF-216-16-250UG AF-216-16-500UG AF-216-16-1MG
10778-490EA 286.72 CAD
10778-490 10778-492 10778-494 10778-496 10778-498 10778-500 10778-502
Mouse Recombinant IL-6 (from E. coli)
Proteins and Peptides
IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6Rα and gp130. Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells. Recombinant Murine IL-6 is a 21.7 kDa protein containing 188 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.