Human Recombinant IL-16 (from E. coli)
Supplier: Peprotech
AF-200-16A-2UG AF-200-16A-10UG AF-200-16A-100UG AF-200-16A-250UG AF-200-16A-500UG AF-200-16A-1MG
10778-346EA 289.23 CAD
10778-346 10778-348 10778-350 10778-352 10778-354 10778-356
Human Recombinant IL-16 (from E. coli)
Proteins and Peptides
IL-16 is a CD8+ T cell-derived cytokine that induces chemotaxis of CD4+ T cells, CD4+ monocytes, and eosinophils. Analysis by gel filtration suggests that, under physiological conditions, hIL-16 exists predominantly as a noncovalently linked multimer, but that some IL-16 may exist as a monomer. However, only the multimeric form appears to possess chemotactic activity, suggesting that receptor cross-linking may be required for activity. IL-16 also induces expression of IL-2 receptor (IL-2R) and MHC class II molecules on CD4 + T cells. Human and murine IL-16 show significant cross-species reactivity. Recombinant Human IL-16 is a 12.4 kDa protein consisting of 121 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.