Human Recombinant IL2 (from E. coli)
Supplier: Peprotech
AF-200-02-10UG AF-200-02-50UG AF-200-02-100UG AF-200-02-250UG AF-200-02-500UG AF-200-02-1MG
10778-240EA 311.84 CAD
10778-240 10778-242 10778-244 10778-246 10778-248 10778-250
Human Recombinant IL2 (from E. coli)
Proteins and Peptides
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation.  IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response.  IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes.  Recombinant Human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond. Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFAQSII STLT
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.