Human Recombinant BMP-4 (from E. coli)
Supplier: Peprotech
AF-120-05ET-2UG AF-120-05ET-10UG AF-120-05ET-50UG AF-120-05ET-100UG AF-120-05ET-250UG AF-120-05ET-500UG AF-120-05ET-1MG
10778-158EA 301.95 CAD
10778-158 10778-160 10778-162 10778-164 10778-166 10778-168 10778-170
Human Recombinant BMP-4 (from E. coli)
Proteins and Peptides
Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-β superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body, and are involved in diverse physiological processes during both pre- and postnatal life. Like BMP-7, BMP-4 is involved in the development and maintenance of bone and cartilage. Reduced expression of BMP-4 is associated with a number of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. PeproTech’s
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.