Human Recombinant BMP2 (from E. coli)
Supplier: Peprotech
AF-120-02-2UG AF-120-02-10UG AF-120-02-50UG AF-120-02-100UG AF-120-02-250UG AF-120-02-500UG AF-120-02-1MG
10778-144EA 301.95 CAD
10778-144 10778-146 10778-148 10778-150 10778-152 10778-154 10778-156
Human Recombinant BMP2 (from E. coli)
Proteins and Peptides
BMPs (Bone Morphogenetic Proteins) belong to the TGF-β superfamily of structurally related signaling proteins. BMP-2 is a potent osteoinductive cytokine, capable of inducing bone and cartilage formation in association with osteoconductive carriers such as collagen and synthetic hydroxyapatite. In addition to its osteogenic activity, BMP-2 plays an important role in cardiac morphogenesis, and is expressed in a variety of tissues, including lung, spleen, brain, liver, prostate ovary and small intestine. The functional form of BMP-2 is a 26 kDa protein composed of two identical 114 amino acid polypeptide chains linked by a single disulfide bond. Each BMP-2 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 23 amino acid signal sequence for secretion, and a 259 amino acid propeptide. After dimerization of this precursor, the covalent bonds between the propeptide (which is also a disulfide-linked homodimer) and the mature BMP-2 ligand are cleaved by a furin-type protease. Recombinant Human BMP-2 is a 26.0 kDa homodimeric protein consisting of two 115 amino acid polypeptide chains. Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.